Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol
Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol
Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol
Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol
Thermo Scientific Chemicals
Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol
CAS: 75-50-3 | C3H9N | 59.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number FSA270442500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS75-50-3, 64-17-5
Specifications
Chemical Name or MaterialTrimethylamine
CAS75-50-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Highly flammable liquid and vapor.
CAS64-17-5
View more
For Manufacturing and Laboratory Use. Not for beverage use