1-Naphthoyl chloride, 99%
1-Naphthoyl chloride, 99%
1-Naphthoyl chloride, 99%
1-Naphthoyl chloride, 99%
Thermo Scientific Chemicals
1-Naphthoyl chloride, 99%
CAS: 879-18-5 | C11H7ClO | 190.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA270380100
Price (MYR)
389.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
389.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Naphthoyl chloride
CAS879-18-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0