Hydroxylamine hydrochloride, 99+%
Hydroxylamine hydrochloride, 99+%
Hydroxylamine hydrochloride, 99+%
Hydroxylamine hydrochloride, 99+%
Thermo Scientific Chemicals
Hydroxylamine hydrochloride, 99+%
CAS: 5470-11-1 | ClH4NO | 69.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA270102500
Price (MYR)
390.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
390.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHydroxylamine hydrochloride
Melting Point155°C to 158°C
CAS5470-11-1
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
Reduction reagent
RUO – Research Use Only