Bis(2-ethylhexyl) sebacate, 97%
Bis(2-ethylhexyl) sebacate, 97%
Bis(2-ethylhexyl) sebacate, 97%
Bis(2-ethylhexyl) sebacate, 97%
Thermo Scientific Chemicals
Bis(2-ethylhexyl) sebacate, 97%
CAS: 122-62-3 | C26H50O4 | 426.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Plastic drum
Catalog number FSA269920100
Price (MYR)
3,494.00
EA
Add to cart
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
3,494.00
EA
Add to cart
Specifications
Chemical Name or MaterialBis(2-ethylhexyl) sebacate
Acidity0.15mg KOH/g max.
CAS122-62-3
Melting Point-55°C
ColorYellow