Ethyl 3,3-diethoxypropanoate, 98%
Ethyl 3,3-diethoxypropanoate, 98%
Ethyl 3,3-diethoxypropanoate, 98%
Ethyl 3,3-diethoxypropanoate, 98%
Thermo Scientific Chemicals
Ethyl 3,3-diethoxypropanoate, 98%
CAS: 10601-80-6 | C9H18O4 | 190.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA269640250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl 3, 3-diethoxypropanoate
CAS10601-80-6
ColorColorless
Density0.9700g/mL
EINECS Number234-223-1