1-Pentene, 97%
1-Pentene, 97%
1-Pentene, 97%
Thermo Scientific Chemicals
1-Pentene, 97%
CAS: 109-67-1 | C5H10 | 70.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA268665000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Pentene
CAS109-67-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0