Tetrahydrofuran, 99.8%, for HPLC, unstabilized
Tetrahydrofuran, 99.8%, for HPLC, unstabilized
Tetrahydrofuran, 99.8%, for HPLC, unstabilized
Tetrahydrofuran, 99.8%, for HPLC, unstabilized
Thermo Scientific Chemicals
Tetrahydrofuran, 99.8%, for HPLC, unstabilized
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FSA268290025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrahydrofuran
Absorbance(1 cm cell vs water), 0.02 max. at 280nm, 0.04 max. at 270nm, 0.05 max. at 260nm, 0.1 max. at 250nm, 0.26 max. at 240nm, 0.5 max. at 230nm
CAS109-99-9
Health Hazard 1Danger
Acidity0.0002 meq/g max.
View more
For HPLC
RUO – Research Use Only