Chlortetracycline hydrochloride, can be used as secondary standard
Chlortetracycline hydrochloride, can be used as secondary standard
Chlortetracycline hydrochloride, can be used as secondary standard
Chlortetracycline hydrochloride, can be used as secondary standard
Thermo Scientific Chemicals
Chlortetracycline hydrochloride, can be used as secondary standard
Chlortetracycline hydrochloride, CAS # 64-72-2, is a broad-spectrum antibiotic of the tetracycline family
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
Catalog number FSA268245000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialChlortetracycline hydrochloride
TypeChlortetracycline Hydrochloride
Physical FormFine Crystalline Powder
CAS64-72-2
Melting Point220.0°C
View more
General Description
  • Chlortetracycline hydrochloride is a broad-spectrum antibiotic of the tetracycline family
  • Receives its name from a 7-chloro substitution on one of the rings of its parent structure, tetracycline
  • A bacteriostatic that inhibits the 30S ribosomal subunit and prevents peptide formation
  • Can work as an antiprotozoal and antifungal
Applications
  • Chlortetracycline is suitable for use in biochemistry, microbiology, in vivo research, and cell culture
  • Intended for research use only
RUO – Research Use Only
General References:
  1. Chen, C.; Ray, P.; Knowlton, K.F.; Pruden, A.; Xia, K. Effect of composting and soil type on dissipation of veterinary antibiotics in land-applied manures. Chemosphere. 2018, 196:270-279.
  2. Weinroth, M.D.; Scott, H.M.; Norby, B.; Loneragan, G.H.; Noyes, N.R.; Rovira, P.; Doster, E.; Yang, X.; Woerner, D.R.; Morley, P.S.; Belk, K.E. Effects of Ceftiofur and Chlortetracycline on the Resistomes of Feedlot Cattle. Appl Environ Microbiol. 2018, 84(13), e00610-18.