Gum arabic, powder
Thermo Scientific Chemicals
Gum arabic, powder
CAS: 9000-01-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA258850010
Price (MYR)
711.00
Bottle
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
711.00
Bottle
Specifications
Chemical Name or MaterialGum arabic
ColorBeige-Yellow to White
CAS9000-01-5
Physical FormPowder
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.