Buffer solution pH 10, Carbonate buffer
Buffer solution pH 10, Carbonate buffer
Thermo Scientific Chemicals
Buffer solution pH 10, Carbonate buffer
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA258600010
Price (MYR)
388.00
BOT
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
388.00
BOT
Add to cart
Chemical Identifiers
CAS1332-57-6, 144-55-8, 7732-18-5
Specifications
Chemical Name or MaterialBuffer solution pH 10
Health Hazard 1
ColorColorless
CAS1332-57-6
CAS144-55-8
View more
pH buffer
RUO – Research Use Only