Buffer solution pH 4, Citrate buffer
Buffer solution pH 4, Citrate buffer
Buffer solution pH 4, Citrate buffer
Buffer solution pH 4, Citrate buffer
Thermo Scientific Chemicals
Buffer solution pH 4, Citrate buffer
CAS: 68-04-2 | C8H5KO4
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA258580010
Price (MYR)
341.00
BOT
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
341.00
BOT
Add to cart
Chemical Identifiers
CAS68-04-2, 7647-14-5, 7732-18-5
Specifications
Chemical Name or MaterialBuffer solution pH 4
Name NotePhthalate buffer
CAS68-04-2
ColorColorless
CAS7647-14-5
View more
pH buffer
RUO – Research Use Only