D(-)-Tagatose, 99+%
D(-)-Tagatose, 99+%
D(-)-Tagatose, 99+%
D(-)-Tagatose, 99+%
D(-)-Tagatose, >-99.0%, C6H12O6, CAS Number-87-81-0, tagatose, d, d--tagatose, d-lyxo-hex-2-ulose, 3s,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, unii-t7a20y888y, naturlose, tagatose nf, lyxo-2-hexulose, keto-d-tagatose, 1g, (5% soln. in H2O) clear colorless to almost colorless, 99+%, 01, 4414 | CAS: 87-81-0 | C6H12O6 | 180.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA257892500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD(-)-Tagatose
CAS87-81-0
ColorWhite
Loss on Drying1.0% max.
Infrared SpectrumAuthentic