View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialLauryl chloride
Name Note99%
CAS112-52-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Causes skin irritation. Very toxic to aquatic life with long lasting effects.