3-Cyclohexyl-1-propanol, 99%
3-Cyclohexyl-1-propanol, 99%
3-Cyclohexyl-1-propanol, 99%
3-Cyclohexyl-1-propanol, 99%
Thermo Scientific Chemicals
3-Cyclohexyl-1-propanol, 99%
CAS: 1124-63-6 | C9H18O | 142.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA2558800505 mLGlass bottle
Catalog number FSA255880050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Cyclohexyl-1-propanol
Name Note99%
CAS1124-63-6
CAS Min %98.5
CAS Max %100.0