4-Methyl-2-pentanone, 99.5%, for analysis
4-Methyl-2-pentanone, 99.5%, for analysis
4-Methyl-2-pentanone, 99.5%, for analysis
4-Methyl-2-pentanone, 99.5%, for analysis
Thermo Scientific Chemicals
4-Methyl-2-pentanone, 99.5%, for analysis
CAS: 108-10-1 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA255660025
Price (MYR)
719.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
719.00
EA
Add to cart
Specifications
Chemical Name or Material4-Methyl-2-pentanone
Acidity0.0004 meq/g max.
CAS108-10-1
CAS Min %99.4
Health Hazard 1GHS Signal Word: Danger