4-Methylvaleric acid, 99%
4-Methylvaleric acid, 99%
4-Methylvaleric acid, 99%
Thermo Scientific Chemicals
4-Methylvaleric acid, 99%
CAS: 646-07-1 | C6H12O2 | 116.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA255510050
Price (MYR)
317.00
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
317.00
Each
Specifications
Chemical Name or Material4-Methylvaleric acid
Name Note99%
CAS646-07-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger