Sodium hyaluronate, 95%
Sodium hyaluronate, 95%
Sodium hyaluronate, 95%
Sodium hyaluronate, 95%
Thermo Scientific Chemicals
Sodium hyaluronate, 95%
CAS: 9067-32-7 | (C14H20NO11Na)n | 417.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA251770250
Price (MYR)
4,810.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,810.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium hyaluronate
CAS9067-32-7
ColorWhite to Cream
pH6.0 to 8.0 (0.5% aq. soln.)
Loss on Drying10% max. (500mg, 105°C, 6 hrs)