Methyltrimethoxysilane, 97%
Methyltrimethoxysilane, 97%
Methyltrimethoxysilane, 97%
Methyltrimethoxysilane, 97%
Thermo Scientific Chemicals
Methyltrimethoxysilane, 97%
CAS: 1185-55-3 | C4H12O3Si | 136.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
1 L
Packaging:
Glass bottle
Catalog number FSA250542500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyltrimethoxysilane
Name Note97%
CAS1185-55-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger