3-Iodophenol, 99%
3-Iodophenol, 99%
3-Iodophenol, 99%
Thermo Scientific Chemicals
3-Iodophenol, 99%
CAS: 626-02-8 | C6H5IO | 220 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA249700250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Iodophenol
Name Note99%
CAS626-02-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning