(2S,3S)-(+)-2,3-Butanediol, 99%
(2S,3S)-(+)-2,3-Butanediol, 99%
(2S,3S)-(+)-2,3-Butanediol, 99%
(2S,3S)-(+)-2,3-Butanediol, 99%
Thermo Scientific Chemicals
(2S,3S)-(+)-2,3-Butanediol, 99%
CAS: 19132-06-0 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
Catalog number FSA247730025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 g
Request bulk or custom format
Specifications
Chemical Name or Material(2S, 3S)-(+)-2, 3-Butanediol
Name Note99%
CAS19132-06-0
CAS Min %98.5
Health Hazard 1HYGROSCOPIC
View more
Chiral derivatization reagent
RUO – Research Use Only