2,5-Dihydrofuran, 98%
2,5-Dihydrofuran, 98%
2,5-Dihydrofuran, 98%
2,5-Dihydrofuran, 98%
Thermo Scientific Chemicals
2,5-Dihydrofuran, 98%
CAS: 1708-29-8 | C4H6O | 70.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA244781000
Price (MYR)
1,821.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,821.00
EA
Add to cart
Specifications
Chemical Name or Material2,5-Dihydrofuran
CAS1708-29-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0