Magnesium stearate, 3.8-5.0% Mg
Magnesium stearate, 3.8-5.0% Mg
Magnesium stearate, 3.8-5.0% Mg
Magnesium stearate, 3.8-5.0% Mg
Thermo Scientific Chemicals
Magnesium stearate, 3.8-5.0% Mg
CAS: 557-04-0 | C36H70MgO4 | 591.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA240760250
Price (MYR)
223.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
223.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMagnesium stearate
CAS557-04-0
Melting Point130.0°C to 140.0°C
ColorWhite
EINECS Number209-150-3