2-(Trifluoromethyl)benzaldehyde, 97%
2-(Trifluoromethyl)benzaldehyde, 97%
2-(Trifluoromethyl)benzaldehyde, 97%
2-(Trifluoromethyl)benzaldehyde, 97%
Thermo Scientific Chemicals
2-(Trifluoromethyl)benzaldehyde, 97%
CAS: 447-61-0 | C8H5F3O | 174.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA232681000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-(Trifluoromethyl)benzaldehyde
CAS447-61-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0