2-Thiopheneacetyl chloride, 99%
2-Thiopheneacetyl chloride, 99%
2-Thiopheneacetyl chloride, 99%
2-Thiopheneacetyl chloride, 99%
Thermo Scientific Chemicals
2-Thiopheneacetyl chloride, 99%
CAS: 39098-97-0 | C6H5ClOS | 160.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA232230500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Thiopheneacetyl chloride
Name Note99%
CAS39098-97-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger