4-(Aminomethyl)benzoic acid, 99%
4-(Aminomethyl)benzoic acid, 99%
4-(Aminomethyl)benzoic acid, 99%
4-(Aminomethyl)benzoic acid, 99%
Thermo Scientific Chemicals
4-(Aminomethyl)benzoic acid, 99%
CAS: 56-91-7 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA232220250
Price (MYR)
539.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
539.00
BOT
Add to cart
Specifications
Chemical Name or Material4-(Aminomethyl)benzoic acid
Name Note99%
CAS56-91-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning