Polyvinylpyrrolidone-iodine complex
Polyvinylpyrrolidone-iodine complex
Polyvinylpyrrolidone-iodine complex
Polyvinylpyrrolidone-iodine complex
Thermo Scientific Chemicals
Polyvinylpyrrolidone-iodine complex
CAS: 25655-41-8 | (C6H9NO)nI2 | 364.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Catalog number FSA229475000
Price (MYR)
1,914.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,914.00
EA
Add to cart
Specifications
Chemical Name or MaterialPolyvinylpyrrolidone-iodine complex
CAS25655-41-8
ColorBrown-Red to Yellow
RTECS NumberTR1579600
TSCATSCA