Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
Thermo Scientific Chemicals
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
1 kg
Packaging:
HDPE plastic bottle
Catalog number FSA228345000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBrij™ 35
ColorWhite
Acid Value5mg KOH/g max.
CAS9002-92-0
Physical FormWaxy Pellets