Pyridoxal 5-phosphate monohydrate, 99%
Pyridoxal 5-phosphate monohydrate, 99%
Pyridoxal 5-phosphate monohydrate, 99%
Pyridoxal 5-phosphate monohydrate, 99%
Thermo Scientific Chemicals
Pyridoxal 5-phosphate monohydrate, 99%
CAS: 41468-25-1 | C8H10NO6P | 247.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA228170250
Price (MYR)
2,543.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,543.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyridoxal 5-phosphate monohydrate
CAS41468-25-1
Melting Point140.0°C to 143.0°C
ColorYellow
Infrared SpectrumAuthentic