Digoxin, 95%
Digoxin, 95%
Digoxin, 95%
Thermo Scientific Chemicals
Digoxin, 95%
CAS: 20830-75-5 | C41H64O14 | 780.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA228120010
Price (MYR)
898.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
898.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDigoxin
CAS20830-75-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal if swallowed.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
R