Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
CAS: 1185-53-1 | C4H11NO3·HCl | 157.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA228032500
Price (MYR)
551.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
551.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTris(hydroxymethyl)aminomethane hydrochloride
CAS1185-53-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Melting Point150.0°C to 153.0°C
View more
Biological buffer
RUO – Research Use Only