Chloramine-T trihydrate, 97+%
Chloramine-T trihydrate, 97+%
Chloramine-T trihydrate, 97+%
Chloramine-T trihydrate, 97+%
Thermo Scientific Chemicals
Chloramine-T trihydrate, 97+%
Chloramine-T trihydrate, >-97%, C7H14ClNNaO5S, CAS Number-7080-50-4, 5kg, 97+%, White, 192 deg.C, 281.69, GHS Signal Word: Danger | CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
1 kg
Packaging:
Plastic Bottle
Catalog number FSA227852500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialChloramine-T trihydrate
TypeChloramine T Trihydrate
Physical FormCrystalline Powder
CAS7080-50-4
Health Hazard 1GHS Signal Word: Danger
View more
C-X bond formation reagent, Oxidation reagent
RUO – Research Use Only