L-1-4'-Tosylamino-2-phenylethyl chloromethyl ketone, 99+%
L-1-4'-Tosylamino-2-phenylethyl chloromethyl ketone, 99+%
L-1-4'-Tosylamino-2-phenylethyl chloromethyl ketone, 99+%
L-1-4'-Tosylamino-2-phenylethyl chloromethyl ketone, 99+%
Thermo Scientific Chemicals
L-1-4'-Tosylamino-2-phenylethyl chloromethyl ketone, 99+%
CAS: 402-71-1 | C17H18ClNO3S | 351.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA227800010
Price (MYR)
1,316.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,316.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-1-4′-Tosylamino-2-phenylethyl chloromethyl ketone
CAS402-71-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye damage.
Health Hazard 3GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.