5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure
Thermo Scientific Chemicals
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure
CAS: 7240-90-6 | C14H15BrClNO6 | 408.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
10 mg
100 mg
Packaging:
Glass bottle
Catalog number FSA227775000
Price (MYR)
1,209.00
BOT
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,209.00
BOT
Add to cart
Specifications
Chemical Name or Material5-Bromo-4-chloro-3-indolyl-β-D-galactoside
CAS7240-90-6
CAS Min %98.5
CAS Max %100.0
Melting Point230.0°C