D(-)-Isoascorbic acid, 98%
D(-)-Isoascorbic acid, 98%
D(-)-Isoascorbic acid, 98%
D(-)-Isoascorbic acid, 98%
Thermo Scientific Chemicals
D(-)-Isoascorbic acid, 98%
CAS: 89-65-6 | C6H7NaO6 | 198.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number FSA227275000
Price (MYR)
319.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
319.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD(-)-Isoascorbic acid
Name Note98%
CAS89-65-6
Melting Point167.0°C to 172.0°C
ColorWhite to Yellow