Hexadecyltrimethylammonium Bromide, 99+%
Hexadecyltrimethylammonium Bromide, 99+%
Hexadecyltrimethylammonium Bromide, 99+%
Hexadecyltrimethylammonium Bromide, 99+%
Thermo Scientific Chemicals
Hexadecyltrimethylammonium Bromide, 99+%
CAS: 57-09-0 | C19H42BrN | 364.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
25 g
100 g
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA227165000
Price (MYR)
1,167.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,167.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHexadecyltrimethylammonium Bromide
ColorWhite
CAS57-09-0
Health Hazard 1Warning
Physical FormCrystalline Powder
View more
Phase transfer catalyst
RUO – Research Use Only