D-Serine, 98%
D-Serine, 98%
D-Serine, 98%
Thermo Scientific Chemicals
D-Serine, 98%
CAS: 312-84-5 | C3H7NO3 | 105.093 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass Bottle
Catalog number FSA227070250
Price (MYR)
658.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
658.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD-Serine
Melting Point220.0°C
CAS312-84-5
Assay Percent Range98%
Molecular FormulaC3H7NO3