1-Naphthyl phosphate, monosodium salt monohydrate, 98+%
CAS: 81012-89-7 | C10H7O4P | 222.14 g/mol
Have Questions?
Change view
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA226890250
Price (MYR)
1,863.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,863.00
EA
Add to cart
Specifications
Chemical Name or Material1-Naphthyl phosphate, monosodium salt monohydrate
Name Note98+%
CAS81012-89-7
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning