Methyl behenate, 99%, analytical standard for GC
Methyl behenate, 99%, analytical standard for GC
Methyl behenate, 99%, analytical standard for GC
Methyl behenate, 99%, analytical standard for GC
Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC
CAS: 929-77-1 | C23H46O2 | 354.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA226810050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl behenate
CAS929-77-1
Melting Point54.0°C to 56.0°C
EINECS Number213-207-8
Infrared SpectrumAuthentic