Atropine sulfate monohydrate, 97+%
Atropine sulfate monohydrate, 97+%
Atropine sulfate monohydrate, 97+%
Atropine sulfate monohydrate, 97+%
Thermo Scientific Chemicals
Atropine sulfate monohydrate, 97+%
Atropine sulfate monohydrate, CAS # 5908-99-6, is a muscarinic receptor antagonist. | CAS: 5908-99-6 | H2SO4·H2O | 694.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA226681000
Price (MYR)
3,641.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,641.00
EA
Add to cart
Specifications
Chemical Name or MaterialAtropine sulfate monohydrate
AcidityPasses Test
CAS5908-99-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Fatal if swallowed.
Causes skin irritation.
Fatal if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
View more
General Description
  • Atropine is a competitive and reversible antagonist of muscarinic acetylcholine receptors on both central and peripheral neurons
  • Atropine was originally isolated from the belladonna plant Atropa belladonna but is also found in other Solanaceae plants
  • Atropine is a drug that can be used to slow heart rate, decrease salivary secretions, and treat poisoning caused by cholinergic or muscarinic agents
Applications
  • It is used to block activation of muscarinic receptors in laboratory mice
  • It can be used in a laboratory setting to decrease oral and airway secretions in research animals
  • Atropine has been used to study heart rate and blood pressure variability in swine
RUO – Research Use Only
General References:
  1. Driessen, A.K.; McGovern, A.E.; Behrens, R.; Moe, A.A.K.; Farrell, M.J.; Mazzone, S.B. A role for neurokinin 1 receptor expressing neurons in the paratrigeminal nucleus in bradykinin-evoked cough in guinea-pigs. J Physiol. 2020, 598(11), 2257-2275.
  2. Egorov, A.V.; Schumacher, D.; Medert, R., Birnbaumer, L.; Freichel, M.; Draguhn, A. TRPC channels are not required for graded persistent activity in entorhinal cortex neurons. Hippocampus. 2019 Nov;29(11):1038-1048. doi: 10.1002/hipo.23094.
  3. Poletto, R.; Janczak, A.M.; Marchant-Forde, R.M.; Marchant-Forde, J.N.; Matthews, D.L.; Dowell, C.A.; Hogan, D.F.; Freeman, L.J.; Lay, D.C. Jr. Identification of low and high frequency ranges for heart rate variability and blood pressure variability analyses using pharmacological autonomic blockade with atropine and propranolol in swine. Physiol Behav. 2011 May 3;103(2):188-96.