6-Mercaptopurine monohydrate, 98%
6-Mercaptopurine monohydrate, 98%
6-Mercaptopurine monohydrate, 98%
6-Mercaptopurine monohydrate, 98%
Thermo Scientific Chemicals
6-Mercaptopurine monohydrate, 98%
CAS: 6112-76-1 | C5H6N4OS | 170.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA226520050
Price (MYR)
578.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
578.00
BOT
Add to cart
Specifications
Chemical Name or Material6-Mercaptopurine monohydrate
Name Note99.5+%
CAS6112-76-1
CAS Min %99.5
Health Hazard 1GHS Signal Word: Warning