Dodecyl sulfate, sodium salt, 85%, pure
Dodecyl sulfate, sodium salt, 85%, pure
Dodecyl sulfate, sodium salt, 85%, pure
Dodecyl sulfate, sodium salt, 85%, pure
Thermo Scientific Chemicals
Dodecyl sulfate, sodium salt, 85%, pure
CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA226145000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDodecyl sulfate, sodium salt
ColorWhite to Yellow
CAS151-21-3
Health Hazard 1Danger
Physical FormPowder or Crystals