Bacitracin, potency min. 60 Units/mg
Bacitracin, potency min. 60 Units/mg
Bacitracin, potency min. 60 Units/mg
Bacitracin, potency min. 60 Units/mg
Thermo Scientific Chemicals
Bacitracin, potency min. 60 Units/mg
Bacitracin, CAS # 1405-87-4, is cyclic polypeptide-based antibiotic which exhibits activity against microbes causing pneumonia, skin, and eye infections.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number FSA226100050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBacitracin
TypeBacitracin
Physical FormPowder
CAS1405-87-4
Health Hazard 1Warning
View more
General Description
• Bacitracin is a mixture of at least nine different cyclic polypeptides, originally isolated from Bacillus bacteria in 1945
• Bacitracins work by binding divalent metal cations and preventing the hydrolysis of lipids into usable cell wall components
Applications
• Bacitracin has been paired with nanoparticles in some laboratory studies to increase its effectivity
• It is suitable for the laboratory study of bacterial resistance, infection, and cross-reactivity
RUO – Research Use Only
General References:
  1. Tay, W.M.; Epperson, J.D.; da Silva, G.F.; Ming, L.J. 1H NMR, mechanism, and mononuclear oxidative activity of the antibiotic metallopeptide bacitracin: the role of D-Glu-4, interaction with pyrophosphate moiety, DNA binding and cleavage, and bioactivity. J Am Chem Soc. 2010, 28, 132(16), 5652-61.
  2. Karala, A.R.; Ruddock, L.W. Bacitracin is not a specific inhibitor of protein disulfide isomerase. FEBS J. 2010, 277(11), 2454-62.