Xylitol, 99+%
Xylitol, 99+%
Xylitol, 99+%
Thermo Scientific Chemicals
Xylitol, 99+%
CAS: 87-99-0 | C5H12O5 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA225985000
Price (MYR)
597.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
597.00
EA
Add to cart
Specifications
Chemical Name or MaterialXylitol
Name Note99+%
CAS87-99-0
Health Hazard 1HYGROSCOPIC
Melting Point92.0°C to 96.0°C