L(-)-Sorbose, 98%
L(-)-Sorbose, 98%
L(-)-Sorbose, 98%
Thermo Scientific Chemicals
L(-)-Sorbose, 98%
CAS: 87-79-6 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA225961000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialL(-)-Sorbose
Name Note99%
CAS87-79-6
Melting Point158°C to 160°C