Adenosine 3',5'-cyclic monophosphate, 99+%
Adenosine 3',5'-cyclic monophosphate, 99+%
Adenosine 3',5'-cyclic monophosphate, 99+%
Adenosine 3',5'-cyclic monophosphate, 99+%
Thermo Scientific Chemicals
Adenosine 3',5'-cyclic monophosphate, 99+%
CAS: 60-92-4 | C10H12N5O6P | 329.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Catalog number FSA225805000
Price (MYR)
596.00
EA
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
596.00
EA
Add to cart
Specifications
Chemical Name or MaterialAdenosine 3′,5′-cyclic monophosphate
CAS60-92-4
Absorbancelambda max. 260nm,pH 7,0.77 to 0.83 (A250/A260),0.13 to 0.19 (A280/A260)
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger