Potassium, 98%, chunks, in mineral oil
Potassium, 98%, chunks, in mineral oil
Potassium, 98%, chunks, in mineral oil
Potassium, 98%, chunks, in mineral oil
Thermo Scientific Chemicals
Potassium, 98%, chunks, in mineral oil
CAS: 7440-09-7 | K | 39.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
50 g
Packaging:
View all
Plastic bottle
Catalog number FSA223861000
Price (MYR)
1,314.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,314.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPotassium
ColorGray
CAS7440-09-7
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.