Antimony(III) sulfide, 98%
Antimony(III) sulfide, 98%
Antimony(III) sulfide, 98%
Antimony(III) sulfide, 98%
Thermo Scientific Chemicals
Antimony(III) sulfide, 98%
CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA223591000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point1150°C
Chemical Name or MaterialAntimony(III) sulfide
Melting Point550°C
Name Note98%
CAS1345-04-6