Perchloric acid, for analysis, ca. 70% solution in water
Perchloric acid, for analysis, ca. 70% solution in water
Perchloric acid, for analysis, ca. 70% solution in water
Thermo Scientific Chemicals
Perchloric acid, for analysis, ca. 70% solution in water
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA223312500
Price (MYR)
2,655.00
Bottle
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,655.00
Bottle
Specifications
Chemical Name or MaterialPerchloric acid
Name Notep.a.
CAS7601-90-3
Melting Point-18.0°C
Boiling Point203.0°C
View more
Oxidation reagent for analysis
RUO – Research Use Only