Chromium potassium sulfate dodecahydrate, 98+%
Chromium potassium sulfate dodecahydrate, 98+%
Chromium potassium sulfate dodecahydrate, 98+%
Chromium potassium sulfate dodecahydrate, 98+%
Thermo Scientific Chemicals
Chromium potassium sulfate dodecahydrate, 98+%
CAS: 7788-99-0 | CrKO8S2·12H2O | 499.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA222525000
Price (MYR)
806.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
806.00
BOT
Add to cart
Specifications
Chemical Name or MaterialChromium potassium sulfate dodecahydrate
Melting Point89.0°C
Name Notep.a.
CAS7788-99-0
ColorPurple