Pyridine hydrochloride, 98%, pure
Pyridine hydrochloride, 98%, pure
Pyridine hydrochloride, 98%, pure
Pyridine hydrochloride, 98%, pure
Thermo Scientific Chemicals
Pyridine hydrochloride, 98%, pure
CAS: 628-13-7 | C5H5N·ClH | 115.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA222285000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPyridine hydrochloride
CAS628-13-7
Health Hazard 1GHS Signal Word: Warning
Melting Point140.0°C to 146.0°C
ColorTan to White